Learn More
Apexbio Technology LLC Rucaparib (free base) 283173-50-2 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062123
Encompass_Preferred
Rucaparib (free base) (CAS 283173-50-2) also known as AG014699 or PF-01367338 is a pharmacological inhibitor targeting poly(ADP-ribose) polymerase (PARP) a nuclear enzyme centrally involved in DNA damage signaling and base excision repair pathways By impeding PARP activity Rucaparib compromises DNA repair capacity leading to persistent DNA strand breaks as evidenced by increased gamma-H2AX and p53BP1 foci It has demonstrated radiosensitizing properties in prostate cancer cells notably those harboring PTEN deficiency or expressing ETS gene fusion proteins due partly to inhibition of non-homologous end-joining (NHEJ) DNA repair Rucaparib serves as a tool compound in oncology research to study chemosensitization and radiosensitization strategies
Catalog No. 50-000-62123 Supplier Apexbio Technology LLC Supplier No. A88935.1
Rucaparib (free base) (CAS 283173-50-2) also known as AG014699 or PF-01367338 is a pharmacological inhibitor targeting poly(ADP-ribose) polymerase (PARP) a nuclear enzyme centrally involved in DNA damage signaling and base excision repair pathways By impeding PARP activity Rucaparib compromises DNA repair capacity leading to persistent DNA strand breaks as evidenced by increased gamma-H2AX and p53BP1 foci It has demonstrated radiosensitizing properties in prostate cancer cells notably those harboring PTEN deficiency or expressing ETS gene fusion proteins due partly to inhibition of non-homologous end-joining (NHEJ) DNA repair Rucaparib serves as a tool compound in oncology research to study chemosensitization and radiosensitization strategies