Learn More
Enzo Life Sciences Rucaparib (free base) (5mg). CAS: 283173-50-2
SDP
Catalog No. 502013643
Encompass_Preferred
Rucaparib is a poly (ADP ribose) polymerase (PARP) inhibitor. PARP is a DNA damage-activated nuclear enzyme that has a key signaling role in the base excision repair pathway. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-3643 Supplier Enzo Life Sciences Supplier No. ENZCHM2350005
Rucaparib is a poly (ADP ribose) polymerase (PARP) inhibitor. PARP is a DNA damage-activated nuclear enzyme that has a key signaling role in the base excision repair pathway. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.