Learn More
Medchemexpress LLC Rucaparib monocamsylate | 1859053-21-6 | MFCD31382198 | >=98.0% | 555.66 g/mol | C29H34FN3O5S | 250 MG
SDP
Catalog No. 5000247399
Encompass_Preferred
Rucaparib monocamsylate is the camphorsulfonate (monocamsylate) salt of rucaparib, an orally active, potent inhibitor of poly(ADP-ribose) polymerases (PARP-1, PARP-2, PARP-3). It exhibits low-nanomolar potency (Ki ≈ 1.4 nM for PARP1) and is used in biochemical and cellular research on DNA repair and oncology.
  • Potent PARP inhibitor with low-nanomolar activity.
  • Used in DNA repair and oncology research applications.
  • Provided as the monocamsylate (camsylate) salt.
  • Suitable for biochemical and cellular assays.
  • Characterized by CAS 1859053-21-6 and molecular weight 555.66 g/mol.
Catalog No. 50-002-47399 Supplier Medchemexpress LLC Supplier No. HY102003250MG
Rucaparib monocamsylate is the camphorsulfonate (monocamsylate) salt of rucaparib, an orally active, potent inhibitor of poly(ADP-ribose) polymerases (PARP-1, PARP-2, PARP-3). It exhibits low-nanomolar potency (Ki ≈ 1.4 nM for PARP1) and is used in biochemical and cellular research on DNA repair and oncology.
  • Potent PARP inhibitor with low-nanomolar activity.
  • Used in DNA repair and oncology research applications.
  • Provided as the monocamsylate (camsylate) salt.
  • Suitable for biochemical and cellular assays.
  • Characterized by CAS 1859053-21-6 and molecular weight 555.66 g/mol.