Learn More
TARGETMOL CHEMICALS INC RUCAPARIB PHOSPHATE 10MG
SDP
Catalog No. 502576594
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Rucaparib Phosphate (PF-01367338 phosphate) is an inhibitor of PARP (Ki 1.4 nM for PARP1) that is used in clinical therapy to sensitize cancer cells to chemotherapy. purity: 99%
Catalog No. 50-257-6594 Supplier TARGETMOL CHEMICALS INC Supplier No. T612710MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Rucaparib Phosphate (PF-01367338 phosphate) is an inhibitor of PARP (Ki 1.4 nM for PARP1) that is used in clinical therapy to sensitize cancer cells to chemotherapy. purity: 99%