Learn More
Selleck Chemical LLC Rucaparib phosphate S1098-1g
SDP
Catalog No. 503108477
Encompass_Preferred
Rucaparib phosphate is an inhibitor of PARP with Ki of 1 4 nM for PARP1 in a cell-free assay also showing binding affinity to eight other PARP domains Phase 3
Catalog No. 50-310-8477 Supplier Selleck Chemical LLC Supplier No. S10981G
Rucaparib phosphate is an inhibitor of PARP with Ki of 1 4 nM for PARP1 in a cell-free assay also showing binding affinity to eight other PARP domains Phase 3