Learn More
Medchemexpress LLC HY-10617 200mg , Rucaparib (phosphate) AG-014699 phosphate;PF-01367338 phosphate CAS:459868-92-9 Purity:98%
SDP
Catalog No. 501492534
Encompass_Preferred
Medchemexpress, HY-10617 200mg Rucaparib (phosphate) AG-014699 phosphate;PF-01367338 phosphate CAS:459868-92-9 Formula:C19H21FN3O5P Ki: 1.4 nM (PARP1) Purity:98% Rucaparib (phosphate) is an inhibitor of  PARP  with  K i  of 1.4 nM for PARP1 in a cell-free assay, and also shows binding affinity to eight other PARP domains. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2534 Supplier Medchemexpress LLC Supplier No. HY10617200MG
Medchemexpress, HY-10617 200mg Rucaparib (phosphate) AG-014699 phosphate;PF-01367338 phosphate CAS:459868-92-9 Formula:C19H21FN3O5P Ki: 1.4 nM (PARP1) Purity:98% Rucaparib (phosphate) is an inhibitor of  PARP  with  K i  of 1.4 nM for PARP1 in a cell-free assay, and also shows binding affinity to eight other PARP domains. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.