Learn More
Medchemexpress LLC Rucaparib phosphate | 459868-92-9 | 99.7% | 421.36 g/mol | C19H21FN3O5P | 10 MG
SDP
Catalog No. 5000242693
Encompass_Preferred
Rucaparib phosphate is the phosphate salt of rucaparib, a potent orally active inhibitor of poly(ADP-ribose) polymerases (PARP-1, PARP-2, PARP-3) used in preclinical research. Supplied as a high-purity solid with recommended sealed storage, the product is available in multiple pack sizes including a 10 mg solid and DMSO solutions for assay use.
  • Potent inhibitor of PARP-1, PARP-2, and PARP-3 with low nanomolar activity.
  • High purity suitable for biochemical and cell-based assays.
  • Available as small solid packs and ready-to-use DMSO solutions.
  • Stable when stored sealed and kept away from moisture.
  • Suitable for research on DNA damage response and cancer models.
Catalog No. 50-002-42693 Supplier Medchemexpress LLC Supplier No. HY1061710MG
Rucaparib phosphate is the phosphate salt of rucaparib, a potent orally active inhibitor of poly(ADP-ribose) polymerases (PARP-1, PARP-2, PARP-3) used in preclinical research. Supplied as a high-purity solid with recommended sealed storage, the product is available in multiple pack sizes including a 10 mg solid and DMSO solutions for assay use.
  • Potent inhibitor of PARP-1, PARP-2, and PARP-3 with low nanomolar activity.
  • High purity suitable for biochemical and cell-based assays.
  • Available as small solid packs and ready-to-use DMSO solutions.
  • Stable when stored sealed and kept away from moisture.
  • Suitable for research on DNA damage response and cancer models.