Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rucaparib phosphate | 459868-92-9 | MFCD17010269 | 98.0% | 421.36 g/mol | C19H21FN3O5P | 100 MG
SDP

Catalog No. 5000244408
Encompass_Preferred

Rucaparib phosphate is the phosphate salt form of rucaparib, a potent poly(ADP-ribose) polymerase (PARP) inhibitor supplied as a research-grade solid for biochemical and cellular studies. The material is characterized by a certificate of analysis and should be stored under conditions specified in the certificate.

  • High purity: 98.0%.
  • Molecular weight: 421.36 g/mol.
  • Chemical formula: C19H21FN3O5P.
  • Appearance: light yellow to yellow solid.
  • Available pack sizes including 100 mg solid.
  • Recommended shipping: room temperature in continental US; storage per certificate of analysis.
  • Intended use: research applications involving PARP inhibition.

Catalog No. 50-002-44408 Supplier Medchemexpress LLC Supplier No. HY10617100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rucaparib phosphate is the phosphate salt form of rucaparib, a potent poly(ADP-ribose) polymerase (PARP) inhibitor supplied as a research-grade solid for biochemical and cellular studies. The material is characterized by a certificate of analysis and should be stored under conditions specified in the certificate.

  • High purity: 98.0%.
  • Molecular weight: 421.36 g/mol.
  • Chemical formula: C19H21FN3O5P.
  • Appearance: light yellow to yellow solid.
  • Available pack sizes including 100 mg solid.
  • Recommended shipping: room temperature in continental US; storage per certificate of analysis.
  • Intended use: research applications involving PARP inhibition.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.