Learn More
Medchemexpress LLC Rucaparib phosphate | 459868-92-9 | MFCD17010269 | 99.8% | 421.4 g/mol | C19H21FN3O5P | 1 ML
SDP
Catalog No. 5000242915
Encompass_Preferred
Rucaparib phosphate is the phosphate salt of rucaparib, a potent poly(ADP-ribose) polymerase (PARP) inhibitor supplied as a 10 mM solution in DMSO (1 mL) for laboratory research applications, particularly in studies of DNA repair and cancer pharmacology.
  • Potent PARP inhibitor activity with low-nanomolar binding affinity.
  • Supplied as a ready-to-use 10 mM solution in DMSO for convenient dosing.
  • High chemical purity (reported >99.7%), suitable for analytical and biological assays.
  • Useful in studies of DNA damage response, synthetic lethality, and oncology models.
  • Phosphate salt improves solubility and handling compared with the free base.
Catalog No. 50-002-42915 Supplier Medchemexpress LLC Supplier No. HY1061710MM 1ML
Rucaparib phosphate is the phosphate salt of rucaparib, a potent poly(ADP-ribose) polymerase (PARP) inhibitor supplied as a 10 mM solution in DMSO (1 mL) for laboratory research applications, particularly in studies of DNA repair and cancer pharmacology.
  • Potent PARP inhibitor activity with low-nanomolar binding affinity.
  • Supplied as a ready-to-use 10 mM solution in DMSO for convenient dosing.
  • High chemical purity (reported >99.7%), suitable for analytical and biological assays.
  • Useful in studies of DNA damage response, synthetic lethality, and oncology models.
  • Phosphate salt improves solubility and handling compared with the free base.