Learn More
Medchemexpress LLC Rucaparib phosphate 50mg | 459868-92-9 | 50MG
SDP
Catalog No. 5000242916
Encompass_Preferred
Rucaparib (AG014699) phosphate is an orally active potent inhibitor of PARP proteins (PARP-1 PARP-2 and PARP-3) with a Ki of 1 4 nM for PARP1 Rucaparib phosphate is a modest hexose-6-phosphate dehydrogenase (H6PD) inhibitor Rucaparib phosphate has the potential for castration-resistant prostate cancer (CRPC) research[1 [2 [3 [4
Catalog No. 50-002-42916 Supplier Medchemexpress LLC Supplier No. HY1061750MG
Rucaparib (AG014699) phosphate is an orally active potent inhibitor of PARP proteins (PARP-1 PARP-2 and PARP-3) with a Ki of 1 4 nM for PARP1 Rucaparib phosphate is a modest hexose-6-phosphate dehydrogenase (H6PD) inhibitor Rucaparib phosphate has the potential for castration-resistant prostate cancer (CRPC) research[1 [2 [3 [4