Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich ™-[(Rucl(Binap))2(Mu-Cl)3[Nh2Me2]

Catalog No. 692433100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-243-3100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 69-243-3100MG Supplier Sigma Aldrich Supplier No. 692433100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Takasago Ligands and Complexes for Asymmetric ReactionsSold in collaboration with Takasago for reseach purposes only. JP Application No. 19960232650

Specifications

CAS 944451-33-6
Molecular Formula C84H114Cl2O8P2Ru
Linear Formula [C84H114ClO8P2Ru]+Cl-
MDL Number MFCD09753019
Quantity 100 mg
Synonym Chloro[(S)-(+)-5,5'-bis[di(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-4,4'-bi-1,3-benzodioxole](p-cymene)ruthenium(II) chloride
Molecular Weight (g/mol) 1485.72 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.