Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[RuCl(p-cymene)((R)-dm-segphos(regR))]Cl, TCI America™
SDP

Catalog No. R0156200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R0156200MG 200 mg
R01561G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R0156200MG Supplier TCI America Supplier No. R0156200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 944451-30-3
Molecular Formula C56H58Cl2O4P2Ru
Molecular Weight (g/mol) 1028.99
MDL Number MFCD09753018
InChI Key LLNJPFQWQJQPEH-UHFFFAOYSA-L
Synonym Chloro[(R)-(+)-5,5′C-bis[di(3,5-xylyl)phosphino]-4,4′C-bi-1,3-benzodioxole](p-cymene)ruthenium(II) Chloride
PubChem CID 131674771
IUPAC Name (4-{5-[bis(3,5-dimethylphenyl)phosphanyl]-2H-1,3-benzodioxol-4-yl}-2H-1,3-benzodioxol-5-yl)bis(3,5-dimethylphenyl)phosphane; 1-methyl-4-(propan-2-yl)benzene; dichlororuthenium
SMILES Cl[Ru]Cl.CC(C)C1=CC=C(C)C=C1.CC1=CC(=CC(C)=C1)P(C1=CC(C)=CC(C)=C1)C1=C(C2=C(OCO2)C=C1)C1=C(C=CC2=C1OCO2)P(C1=CC(C)=CC(C)=C1)C1=CC(C)=CC(C)=C1

Specifications

Quantity 200 mg
Formula Weight 1028.99
Physical Form Crystalline Powder
Chemical Name or Material [RuCl(p-cymene)((R)-dm-segphos(regR))]Cl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.