Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC RuCl(p-cymene)[(R,R)-Ts-DPEN] | 192139-92-7 | 636.21 | 1 G
SDP

Catalog No. 5000449745
Encompass_Preferred

RuCl(p-cymene)[(R,R)-Ts-DPEN] is a biochemical reagent suitable for various life science related research applications. It serves as a biological material or an organic compound, commonly appearing as a yellow to brown solid or powder.

  • Can be used as a biochemical reagent
  • Suitable for life science related research
  • Store at -20°C under nitrogen
  • For solutions, store at -80°C for 6 months or -20°C for 1 month under nitrogen
  • Purity of ≥98%
  • Molecular weight of 636.21
  • CAS number: 192139-92-7

Catalog No. 50-004-49745 Supplier Medchemexpress LLC Supplier No. HYW0010691G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RuCl(p-cymene)[(R,R)-Ts-DPEN] is a biochemical reagent suitable for various life science related research applications. It serves as a biological material or an organic compound, commonly appearing as a yellow to brown solid or powder.

  • Can be used as a biochemical reagent
  • Suitable for life science related research
  • Store at -20°C under nitrogen
  • For solutions, store at -80°C for 6 months or -20°C for 1 month under nitrogen
  • Purity of ≥98%
  • Molecular weight of 636.21
  • CAS number: 192139-92-7

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.