Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Rucl2[™-Dm-Binap][(R,R)-Dpen]

Catalog No. 69230150MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-230-150MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 69-230-150MG Supplier Sigma Aldrich Supplier No. 69230150MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

The product is a bis-naphthalene containing phosphine ligand, in conjugation with a ruthenium complex.Takasago Ligands and Complexes for Asymmetric ReactionsSold in collaboration with Takasago for research purposes only. JP Registration No. 2041996, 2731377, 2935453 JP Application No. 19970359654

Specifications

CAS 566940-03-2
Molecular Formula C74H100O8P2
Linear Formula C74H100O8P2
MDL Number MFCD09753003
Quantity 100 mg
Synonym (R)-(-)-5,5'-Bis[di(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-4,4'-bi-1,3-benzodioxole; [(4R)-(4,4'-bi-1,3-benzodioxole)-5,5'-diyl]bis[bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphine]
Molecular Weight (g/mol) 1179.53 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.