Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RuCl2[(R)-xylbinap][(R)-daipen], TCI America™
SDP

Catalog No. R0133200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R0133200MG 200 mg
R01331G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R0133200MG Supplier TCI America Supplier No. R0133200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 220114-32-9
Molecular Formula C71H74Cl2N2O2P2Ru
MDL Number MFCD09753026

Specifications

Quantity 200 mg
Formula Weight 1221.30
Physical Form Crystalline Powder
Chemical Name or Material RuCl2[(R)-xylbinap][(R)-daipen]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.