Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RuCl2[(S)-dm-segphos(regR)][(S,S)-dpen], TCI America™
SDP

Catalog No. R01301G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R01301G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R01301G Supplier TCI America Supplier No. R01301G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 944450-46-8
Molecular Formula C60H60Cl2N2O4P2Ru
Quantity 1 g
MDL Number MFCD09753032
InChI Key FOEKPXQPRAIJRW-UHFFFAOYNA-L
SMILES Cl[Ru]Cl.NC(C(N)C1=CC=CC=C1)C1=CC=CC=C1.CC1=CC(=CC(C)=C1)P(C1=CC(C)=CC(C)=C1)C1=C(C2=C(OCO2)C=C1)C1=C(C=CC2=C1OCO2)P(C1=CC(C)=CC(C)=C1)C1=CC(C)=CC(C)=C1
IUPAC Name (4-{5-[bis(3,5-dimethylphenyl)phosphanyl]-2H-1,3-benzodioxol-4-yl}-2H-1,3-benzodioxol-5-yl)bis(3,5-dimethylphenyl)phosphane; 1,2-diphenylethane-1,2-diamine; dichlororuthenium
Molecular Weight (g/mol) 1107.07
PubChem CID 71310551
Formula Weight 1107.07
Physical Form Crystalline Powder
Chemical Name or Material RuCl2[(S)-dm-segphos(regR)][(S,S)-dpen]
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.