Learn More
Medchemexpress LLC Rufloxacin hydrochloride | 106017-08-7 | 99.25% | 100 MG
SDP
Catalog No. 5000409343
Encompass_Preferred
Rufloxacin hydrochloride is a fluoroquinolone antibacterial that inhibits B-cell differentiation in human mononuclear cells and inhibits Topo. It is for research use only.
  • Fluoroquinolone antibacterial
  • Inhibits B-cell differentiation in human mononuclear cells
  • Inhibits Topo
  • Solid appearance
  • Light yellow to yellow color
Catalog No. 50-004-09343 Supplier Medchemexpress LLC Supplier No. HYB0902A100MG
Rufloxacin hydrochloride is a fluoroquinolone antibacterial that inhibits B-cell differentiation in human mononuclear cells and inhibits Topo. It is for research use only.
  • Fluoroquinolone antibacterial
  • Inhibits B-cell differentiation in human mononuclear cells
  • Inhibits Topo
  • Solid appearance
  • Light yellow to yellow color