Learn More
Enzo Life Sciences Rufloxacin . hydrochloride (250mg). CAS: 106017-08-7
SDP
Catalog No. 502009023
Encompass_Preferred
Fluoroquinolone antibacterial agent. Structurally similar to ofloxacin. Specific inhibitor of bacterial DNA gyrase (topoisomerase). Inhibits B-cell differentiation. Shows photosensitizing effects. Alternative name: 6-Fluoro-2,3-dihydro-10-(4-methyl-1-piperazinyl)-7-oxo-7H-pyrido[1,2,3-de]-1,4-benzothia zine-6-carboxylic acid . hydrochloride, MF-934 . hydrochloride. Purity: ≥98% (Assay). Appearance: Off-white to light green crystalline solid. Solubility: Sparingly soluble in water; very slightly soluble in methanol; almost insoluble in chloroform or acetic acid. Long Term Storage: -20°C.
Catalog No. 50-200-9023 Supplier Enzo Life Sciences Supplier No. ALX380300M250
Fluoroquinolone antibacterial agent. Structurally similar to ofloxacin. Specific inhibitor of bacterial DNA gyrase (topoisomerase). Inhibits B-cell differentiation. Shows photosensitizing effects. Alternative name: 6-Fluoro-2,3-dihydro-10-(4-methyl-1-piperazinyl)-7-oxo-7H-pyrido[1,2,3-de]-1,4-benzothia zine-6-carboxylic acid . hydrochloride, MF-934 . hydrochloride. Purity: ≥98% (Assay). Appearance: Off-white to light green crystalline solid. Solubility: Sparingly soluble in water; very slightly soluble in methanol; almost insoluble in chloroform or acetic acid. Long Term Storage: -20°C.