Learn More
Glas Col LLC Rotator
SDP
Catalog No. NC0099250
Encompass_Preferred
Rugged Rotator base, with friction-drive test tube head, .17-8.3 rpm, 120V. Furnished with 14" friction drive test tube head. 31 spring clips for 10-18mm diameter tubes and 16 spring clips for 20-30mm diameter tubes.
Catalog No. NC0099250 Supplier Glas Col LLC Supplier No. 099A RD5512
Rugged Rotator base, with friction-drive test tube head, .17-8.3 rpm, 120V. Furnished with 14" friction drive test tube head. 31 spring clips for 10-18mm diameter tubes and 16 spring clips for 20-30mm diameter tubes.