Learn More
Bioworld Rugini Olive Medium, 50 L
Catalog No. 50254777
Encompass_Preferred
Rugini Olive MediumContains Macronutrients, Micronutrients, and Vitamins Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture tested
Catalog No. 50-254-777 Supplier Bioworld Supplier No. 306301104
Rugini Olive MediumContains Macronutrients, Micronutrients, and Vitamins Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture tested