Learn More
AdipoGen Rugulosin (250 µg)
SDP
Catalog No. 502053493
Encompass_Preferred
Chemical. CAS: 23537-16-8. Formula: C30H22O10. MW: 542.5. Isolated from Penicillium sp. Antibiotic. Antibacteriophage. Antiviral. Mycotoxin. DNA replication, transcription and repair inhibitor. Antibacterial. RNA polymerase and ribonuclease H inhibitor. Insecticidal. Cytotoxic. HIV-1 integrase inhibitor. Shows anti-MRSA (methicillin-resistant Staphylococcus aureus) activity. Potential Hsp90 inhibitor through interacting with N-Hsp90.
Catalog No. 50-205-3493 Supplier AdipoGen Supplier No. AGCN20124C250
Chemical. CAS: 23537-16-8. Formula: C30H22O10. MW: 542.5. Isolated from Penicillium sp. Antibiotic. Antibacteriophage. Antiviral. Mycotoxin. DNA replication, transcription and repair inhibitor. Antibacterial. RNA polymerase and ribonuclease H inhibitor. Insecticidal. Cytotoxic. HIV-1 integrase inhibitor. Shows anti-MRSA (methicillin-resistant Staphylococcus aureus) activity. Potential Hsp90 inhibitor through interacting with N-Hsp90.