Learn More
Abcam Rugulosin ((+)-Rugulosin), RNA polymerase inhibitor, 250UG
Catalog No. 502705849
Encompass_Preferred
MW 542.5 Da, Purity >95%. Mycotoxin. RNA polymerase inhibitor. Shows weak hepatocarcinogenic effects in vivo. Shows antibacterial and insecticidal effects. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5849 Supplier Abcam Supplier No. AB144351250UG
MW 542.5 Da, Purity >95%. Mycotoxin. RNA polymerase inhibitor. Shows weak hepatocarcinogenic effects in vivo. Shows antibacterial and insecticidal effects. Orally active.
The product is subject to the following: Abcam Restricted Use Statement