Learn More
AdipoGen Rugulosin -Rugulosin
SDP
Supplier:  AdipoGen BVT0444M005
Encompass_Preferred
Chemical. CAS 23537-16-8. Formula C30H22O10. MW 542.5. Isolated from Penicillium sp. Mycotoxin. Antibiotic Streptococcus, Corynebacterium, MRSA. Antiviral, HIV-1 integrase inhibitor. DNA replication, transcription and repair inhibitor carcinogenic activity. RNA polymerase and ribonuclease H inhibitor. Insecticidal activity. Inhibitor of N-Hsp90.
Catalog No. 50-256-1921