Learn More
Chemglass Life Sciences Ruler, 12", Stainless Steel, Flexible
SDP
Catalog No. 501531367
Encompass_Preferred
Ruler, 12", Stainless Steel , Flexible. Flexible Stainless Steel ruler with black etched graduations. One side is metric with 0.5 and 1.0mm graduations while the other side is graduated in 1/34 subdivisions. Ruler is 7/8 wide.
Catalog No. 50-153-1367 Supplier Chemglass Life Sciences Supplier No. CG1980R60
Ruler, 12", Stainless Steel , Flexible. Flexible Stainless Steel ruler with black etched graduations. One side is metric with 0.5 and 1.0mm graduations while the other side is graduated in 1/34 subdivisions. Ruler is 7/8 wide.