Learn More
Andwin Scientific 12 in. Two-Sided Ruler, Flexible stainless steel. One side has metric scale graduated in 1mm and 0.5mm divisions and English scale graduated in 1/16 , 1/32 and 1/64 in. divisions
SDP
Catalog No. NC9518793
Encompass_Preferred
12 in. Two-Sided Ruler, Flexible stainless steel. One side has metric scale graduated in 1mm and 0.5mm divisions and English scale graduated in 1/16 , 1/32 and 1/64 in. divisions
Catalog No. NC9518793 Supplier Andwin Scientific Supplier No. 56510014
12 in. Two-Sided Ruler, Flexible stainless steel. One side has metric scale graduated in 1mm and 0.5mm divisions and English scale graduated in 1/16 , 1/32 and 1/64 in. divisions