Learn More
Aviva Systems Biology Corp RUNDC2A Antibody - N-terminal region (ARP47692_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47692_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RUNDC2A. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human RUNDC2A. Alternate names: FLJ12363ApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59640