Learn More
Grainger Runner Cutter 1/16 CR Carbide
Catalog No. 502765766
Encompass_Preferred
The Micro 100 Runner Cutter Carbide is a high-performance end mill designed for high-speed machining of aluminum, brass, copper, and other non-ferrous metals. It features a 16mm diameter, 4 flute design with a TiAlN coating for heat and wear resistance. The helical flutes provide efficient chip evacuation and a smooth cutting action. With a corner radius of 0.4mm, this end mill is well-suited for slotting, side milling, and contouring operations. The Micro 100 Runner delivers accurate results across a wide range of feed rates and depths of cut.
Catalog No. 50-276-5766 Supplier Grainger Supplier No. 16T413
The Micro 100 Runner Cutter Carbide is a high-performance end mill designed for high-speed machining of aluminum, brass, copper, and other non-ferrous metals. It features a 16mm diameter, 4 flute design with a TiAlN coating for heat and wear resistance. The helical flutes provide efficient chip evacuation and a smooth cutting action. With a corner radius of 0.4mm, this end mill is well-suited for slotting, side milling, and contouring operations. The Micro 100 Runner delivers accurate results across a wide range of feed rates and depths of cut.