Learn More
Aviva Systems Biology Corp RUNX1 Antibody - N-terminal region (ARP38073_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38073_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RUNX1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RUNX1. Alternate names: AML1, CBFA2, EVI-1, AMLCR1, PEBP2aB, AML1-EVI-1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56509