Learn More
Aviva Systems Biology Corp RUNX1T1 Antibody - middle region (ARP33996_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33996_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RUNX1T1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RUNX1T1. Alternate names: CDR, ETO, MTG8, AML1T1, ZMYND2, CBFA2T1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55217