Learn More
Aviva Systems Biology Corp Runx1t1 Antibody - N-terminal region (ARP58128_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58128_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Runx1t1. It was validated on Western Blot. . Alternate names: Cbfa2t1h, ETO, MTG8ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-441