Learn More
Aviva Systems Biology Corp RUNX2 Antibody - middle region (ARP36678_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36678_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RUNX2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RUNX2. Alternate names: CCD, AML3, CCD1, OSF2, CBFA1, OSF-2, PEA2aA, PEBP2A1, PEBP2A2, PEBP2aA, PEBP2aA1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56247