Learn More
Aviva Systems Biology Corp RUNX2 Antibody - C-terminal region (ARP38453_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38453_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RUNX2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human RUNX2. Alternate names: AML3, CBFA1, CCD, CCD1, MGC120022, MGC120023, OSF2, PEA2aA, PEBP2A1, PEBP2A2, PEBP2aA, PEBP2aA1, OSF-2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56662