Learn More
CAPTIVATE BIO LLC RUO hPL heparin-required
Catalog No. 5000876343
Encompass_Preferred
PLTMax Human Platelet Lysate research-grade 500mL from Captivate Bio is a growth factor-rich supplement that is a superior alternative to FBS PLTMax is used in the generation of MSCs and T-cells including neurology nephrology gastrointestinal disease wound repair and cardiology US origin
Catalog No. 50-008-76343 Supplier CAPTIVATE BIO LLC Supplier No. CBPLTMAX5R
PLTMax Human Platelet Lysate research-grade 500mL from Captivate Bio is a growth factor-rich supplement that is a superior alternative to FBS PLTMax is used in the generation of MSCs and T-cells including neurology nephrology gastrointestinal disease wound repair and cardiology US origin