Learn More
CAPTIVATE BIO LLC RUO hPL xeno-free
Supplier: CAPTIVATE BIO LLC CBPLTGOLD1R
PLTGold Human Platelet Lysate research-grade 100mL from Captivate Bio is a revolutionary xeno-free heparin-free supplement which provides rapid cell expansion of MSCs and immune cells (T cells and NK cells) PLTGold research-grade is an excellent supplement for use in both 2D and 3D culture systems US origin