Learn More
CAPTIVATE BIO LLC RUO hPL xeno-free
Catalog No. 5000876345
Encompass_Preferred
PLTGold Human Platelet Lysate research-grade 500mL from Captivate Bio is a revolutionary xeno-free heparin-free supplement which provides rapid cell expansion of MSCs and immune cells (T cells and NK cells) PLTGold research-grade is an excellent supplement for use in both 2D and 3D culture systems US origin
Catalog No. 50-008-76345 Supplier CAPTIVATE BIO LLC Supplier No. CBPLTGOLD5R
PLTGold Human Platelet Lysate research-grade 500mL from Captivate Bio is a revolutionary xeno-free heparin-free supplement which provides rapid cell expansion of MSCs and immune cells (T cells and NK cells) PLTGold research-grade is an excellent supplement for use in both 2D and 3D culture systems US origin