Learn More
Apexbio Technology LLC Rupatadine Fumarate 182349-12-8 10mM (in 1mL Ethanol)
SDP
Catalog No. 5000062449
Encompass_Preferred
Rupatadine Fumarate (CAS 182349-12-8) is a small molecule antagonist targeting platelet-activating factor receptor (PAFR) and histamine H1 receptor It exhibits binding affinities (K i) of 550 nM at PAFR and 102 nM at the histamine H1 receptor effectively inhibiting signaling mediated by these pathways Through dual antagonism rupatadine fumarate disrupts inflammatory and allergic responses regulated by platelet-activating factor and histamine signaling Currently in Phase 2 clinical development it serves as a valuable pharmacological tool for studying inflammation and allergic conditions
Catalog No. 50-000-62449 Supplier Apexbio Technology LLC Supplier No. B15665.1
Rupatadine Fumarate (CAS 182349-12-8) is a small molecule antagonist targeting platelet-activating factor receptor (PAFR) and histamine H1 receptor It exhibits binding affinities (K i) of 550 nM at PAFR and 102 nM at the histamine H1 receptor effectively inhibiting signaling mediated by these pathways Through dual antagonism rupatadine fumarate disrupts inflammatory and allergic responses regulated by platelet-activating factor and histamine signaling Currently in Phase 2 clinical development it serves as a valuable pharmacological tool for studying inflammation and allergic conditions