Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rupatadine fumarate | 182349-12-8 | MFCD00926499 | 97.3% | C30H30ClN3O4 | 10MG
SDP

Catalog No. 5000259284
Encompass_Preferred

Rupatadine fumarate is an analytical reference standard supplied as a 10 mg pack for research and analytical applications. It corresponds to CAS 182349-12-8, has a molecular formula of C30H30ClN3O4, a molecular weight of 532.03, and is provided with a stated purity of approximately 97.3%. For laboratory research use only.

  • Intended for analytical, method development, and validation use.
  • Available in a 10 mg pack size.
  • Molecular formula C30H30ClN3O4 and molecular weight 532.03.
  • Listed purity 97.3%.
  • Provided for laboratory research use only.

Catalog No. 50-002-59284 Supplier Medchemexpress LLC Supplier No. HY13511AR10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rupatadine fumarate is an analytical reference standard supplied as a 10 mg pack for research and analytical applications. It corresponds to CAS 182349-12-8, has a molecular formula of C30H30ClN3O4, a molecular weight of 532.03, and is provided with a stated purity of approximately 97.3%. For laboratory research use only.

  • Intended for analytical, method development, and validation use.
  • Available in a 10 mg pack size.
  • Molecular formula C30H30ClN3O4 and molecular weight 532.03.
  • Listed purity 97.3%.
  • Provided for laboratory research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.