Learn More
Apexbio Technology LLC Rupatadine Fumarate, 500mg Cas# 182349-12-8
SDP
Supplier:  Apexbio Technology LLC B1566500
Encompass_Preferred
Rupatadine is an inhibitor of PAFR and histamine (H1) receptor with Ki of 550 nM and 102 nM, respectively. Phase 2. Other sizes are available. Please inqury us for quote.
Catalog No. 50-199-8552