Learn More
Selleck Chemical LLC Rupatadine Fumarate S3052-10mg
SDP
Catalog No. 503109352
Encompass_Preferred
Rupatadine is an inhibitor of PAFR and histamine (H1) receptor with Ki of 550 nM and 102 nM respectively
Catalog No. 50-310-9352 Supplier Selleck Chemical LLC Supplier No. S305210MG
Rupatadine is an inhibitor of PAFR and histamine (H1) receptor with Ki of 550 nM and 102 nM respectively