Learn More
TARGETMOL CHEMICALS INC Rupestonic acid 5MG
SDP
Catalog No. 502880296
Encompass_Preferred
Also available in 1 mg, 2 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Rupestonic acid is a compound in Artemisia rupestris L., a well-known traditional Uighur medicine for the treatment of colds. Rupestonic acid has an anti-influenza virus activity. Purity 98.07%
Catalog No. 50-288-0296 Supplier TARGETMOL CHEMICALS INC Supplier No. TN21665MG
Add to Cart
Add to Cart
Also available in 1 mg, 2 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Rupestonic acid is a compound in Artemisia rupestris L., a well-known traditional Uighur medicine for the treatment of colds. Rupestonic acid has an anti-influenza virus activity. Purity 98.07%