Learn More
Strem, An Ascensus Company CAS# 1445085-77-7. 250mg. Methanesulfonato(2-dicyclohexylphosphino-2',6'-di-i-propoxy-1,1'-biphenyl)(2'-amino-1,1'-biphenyl-2-yl)palladium(II), min. 98% [RuPhos Palladacycle Gen. 3]. MFCD22572684
SDP
Supplier:  Strem, An Ascensus Company 460314250MG
Encompass
CAS# 1445085-77-7. 250mg. Methanesulfonato(2-dicyclohexylphosphino-2',6'-di-i-propoxy-1,1'-biphenyl)(2'-amino-1,1'-biphenyl-2-yl)palladium(II), min. 98% [RuPhos Palladacycle Gen. 3]. MFCD22572684. Molecular Weight: 836.37. Molecular Formula: C43H56NO5PPdS. Color/form: white pwdr. Strem# 46-0314. http://www.strem.com/catalog/v/46-0314/
Catalog No. 50-201-6647