Learn More
Strem, An Ascensus Company RuPhos Palladacycle G4, 98% 1g
SDP
Supplier:  Strem, An Ascensus Company 4603951G
Encompass
CAS# 1599466-85-9. 1g. Methanesulfonato(2-dicyclohexylphosphino-2',6'-di-i-propoxy-1,1'-biphenyl)(2'-methylamino-1,1'-biphenyl-2-yl)palladium(II), min. 98% [RuPhos Palladacycle Gen. 4]. MFCD28411659. Molecular Weight: 850.40. Molecular Formula: C44H58NO5PPdS. Color/form: off-white to tan pwdr. Strem# 46-0395. http://www.strem.com/catalog/v/46-0395/
Catalog No. 50-203-4175