Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ruphos Pd G1 Methyl T-F-Butyl Ether Adduct

Catalog No. 707589250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-758-9250MG 250 mg
70-758-91G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 70-758-9250MG Supplier Sigma Aldrich Supplier No. 707589250MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ruphos Pd G1 Methyl T-F-Butyl Ether Adduct

Specifications

CAS 2361069-97-6
Melting Point 180°C to 200°C
Molecular Formula C38H53ClNO2PPd -+ CH3OC4H9
Linear Formula C38H53ClNO2PPd -+ CH3OC4H9
MDL Number MFCD12545955
Quantity 250 mg
Synonym (RuPhos) palladium(II) phenethylamine chloride (1:1 MTBE solvate); Chloro-(2-Dicyclohexylphosphino-2',6'-diisopropoxy-1,1'-biphenyl)[2-(2-aminoethyl)phenyl]palladium(II) - methyl-t-butyl ether adduct; RuPhos Palladacycle; RuPhos precatalyst
Molecular Weight (g/mol) 816.83 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.