Learn More
Strem, An Ascensus Company CAS# 1375325-68-0. 1g. Chloro(2-dicyclohexylphosphino-2',6'-di-i-propoxy-1,1'-biphenyl)(2'-amino-1,1'-biphenyl-2-yl)palladium(II), min. 98% [RuPhos Palladacycle Gen. 2]. MFCD20264899
SDP
Supplier:  Strem, An Ascensus Company 4602861G
Encompass
CAS# 1375325-68-0. 1g. Chloro(2-dicyclohexylphosphino-2',6'-di-i-propoxy-1,1'-biphenyl)(2'-amino-1,1'-biphenyl-2-yl)palladium(II), min. 98% [RuPhos Palladacycle Gen. 2]. MFCD20264899. Molecular Weight: 776.72. Molecular Formula: C42H53ClNO2PPd. Color/form: beige pwdr. Strem# 46-0286. http://www.strem.com/catalog/v/46-0286/
Catalog No. 50-201-6754