Learn More
Medchemexpress LLC HY-106161 1mg Medchemexpress, Rupintrivir CAS:223537-30-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1061611MG
Encompass_Preferred
Medchemexpress, HY-106161 1mg Rupintrivir CAS:223537-30-2 Rupintrivirvr (AG7088), an antiviral drug, is a potent, selective and irreversible inhibitor of human rhinovirus (HRV) 3C protease. Rupintrivirvr inhibits replication of a panel of 48 different HRV serotypes in H1-HeLA and MRC-5 cell protection assays, with a mean EC50 of 0.023 μM. Rupintrivirvr shows immune-modulatory effect[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9563