Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rupintrivir | 223537-30-2 | 99.9% | 598.66 g·mol⁻¹ | C31H39FN4O7 | 25 MG
SDP

Catalog No. 5000242885
Encompass_Preferred

Rupintrivir is a potent, selective, irreversible inhibitor of human rhinovirus (HRV) 3C protease provided as a research-grade small molecule for antiviral and protease inhibition studies.

  • Potent, selective, irreversible inhibition of HRV 3C protease.
  • High purity suitable for biochemical and cellular assays.
  • Molecular weight 598.66 g·mol⁻¹; chemical formula C31H39FN4O7.
  • Available in small powder quantities for research use.
  • Soluble in common organic solvents for stock preparation.

Catalog No. 50-002-42885 Supplier Medchemexpress LLC Supplier No. HY10616125MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rupintrivir is a potent, selective, irreversible inhibitor of human rhinovirus (HRV) 3C protease provided as a research-grade small molecule for antiviral and protease inhibition studies.

  • Potent, selective, irreversible inhibition of HRV 3C protease.
  • High purity suitable for biochemical and cellular assays.
  • Molecular weight 598.66 g·mol⁻¹; chemical formula C31H39FN4O7.
  • Available in small powder quantities for research use.
  • Soluble in common organic solvents for stock preparation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.