Learn More
Welker Inc Welker Burst/Rupture Disc for Constant Pressure Cylinder (CPC)
Supplier:  Welker Inc MR002OO
Encompass
Pressure Rating: 1900 psig ; Tolerance: -7.85% / +11.3% ; Material: ALY600-PTFE ; Torque20 ft. lbs.
Catalog No. NC0237152