Learn More
Braintree Scientific RUSABLE SCRUFF GRD SZ9 20-38GR
SDP
Catalog No. NC1936833
Encompass
With a ScruffGuard, you can handle and dose small animals gently, whilst avoiding bites and needlestick injuries. A ScruffGuard can be used to administer substances by subcutaneous, intraperitoneal and intramuscular routes. It can be used to implant microchips subcutaneously for identification purposes. A ScruffGuard is particularly helpful for trainees who are learning to handle small animals for the first time. Dimensions 16 15 8cm, made from Nylon material. Non-sterile, however the ScruffGuard may be autoclaved at a maximum of 121 degrees celsius.BRAINTREE SCIENTIFIC mfg# SCRUFF-9 **Sold for research purposes only and not for human use**
Catalog No. NC1936833 Supplier Braintree Scientific Supplier No. SCRUFF9
With a ScruffGuard, you can handle and dose small animals gently, whilst avoiding bites and needlestick injuries. A ScruffGuard can be used to administer substances by subcutaneous, intraperitoneal and intramuscular routes. It can be used to implant microchips subcutaneously for identification purposes. A ScruffGuard is particularly helpful for trainees who are learning to handle small animals for the first time. Dimensions 16 15 8cm, made from Nylon material. Non-sterile, however the ScruffGuard may be autoclaved at a maximum of 121 degrees celsius.BRAINTREE SCIENTIFIC mfg# SCRUFF-9 **Sold for research purposes only and not for human use**