Learn More
Medchemexpress LLC HY-105069A 1mg Medchemexpress, Rusalatide acetate CAS:875455-82-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY105069A1MG
Encompass_Preferred
Medchemexpress, HY-105069A 1mg Rusalatide acetate CAS:875455-82-6 Rusalatide acetate (TP508 amide acetate), a regenerative peptide, mitigates radiation-induced gastrointestinal damage by activating stem cells and preserving crypt integrity [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2257