Learn More
Medchemexpress LLC Rusalatide acetate (TP508 amide acetate) | 875455-82-6 | 98.1% | 2371.50 Da | C99H151N29O37S | 10 MG
SDP
Catalog No. 5000243634
Encompass_Preferred
Rusalatide acetate (TP508 amide acetate) is a 23-amino-acid regenerative peptide for research use that mitigates radiation-induced gastrointestinal damage by activating stem cells and preserving crypt integrity. It is supplied as a lyophilized powder intended for experimental applications in tissue regeneration and radioprotection studies.
  • Regenerative peptide that activates stem cells.
  • Mitigates radiation-induced gastrointestinal damage and preserves crypt integrity.
  • Defined 23-amino-acid sequence with amide C-terminus.
  • Reported high purity (about 98.1%).
  • Stable when stored frozen and protected from moisture and light.
Catalog No. 50-002-43634 Supplier Medchemexpress LLC Supplier No. HY105069A10MG
Rusalatide acetate (TP508 amide acetate) is a 23-amino-acid regenerative peptide for research use that mitigates radiation-induced gastrointestinal damage by activating stem cells and preserving crypt integrity. It is supplied as a lyophilized powder intended for experimental applications in tissue regeneration and radioprotection studies.
  • Regenerative peptide that activates stem cells.
  • Mitigates radiation-induced gastrointestinal damage and preserves crypt integrity.
  • Defined 23-amino-acid sequence with amide C-terminus.
  • Reported high purity (about 98.1%).
  • Stable when stored frozen and protected from moisture and light.